Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INPP5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552921
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552921 Supplier Invitrogen™ Supplier No. PA552921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVVKLIFRES DNDRKVMLQL EKKLFDYFNQ EVFRDNNGTA LLEFDKELSV FKDRLYELDI SFPPSYPYSE DARQGEQYMN TRCPAWCDRI LMSPSAKELV LRSESEEKVV TYDHIGPNVC MGDHKPVFL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

The protein encoded by this gene is a membrane-associated type I inositol 1,4,5-trisphosphate (InsP3) 5-phosphatase. InsP3 5-phosphatases hydrolyze Ins(1,4,5)P3, which mobilizes intracellular calcium and acts as a second messenger mediating cell responses to various stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP5A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INPP5A
Gene Accession No. Q14642
Gene Alias 43 kDa inositol polyphosphate 5-phophatase; 43 kDa inositol polyphosphate 5-phosphatase; 5PTase; CTCL tumor antigen HD-CL-02; DKFZp434A1721; inositol polyphosphate-5-phosphatase A; inositol polyphosphate-5-phosphatase, 40kD; inositol polyphosphate-5-phosphatase, 40kDa; inositol trisphosphate-5-phosphatase, 40kD; inositol-1,4,5-trisphosphate 5-phosphatase; Inpp5a; InsP3 5-phosphatase; MGC116947; MGC116949; type I inositol 1,4,5-trisphosphate 5-phosphatase; type I inositol-1,4,5-trisphosphate 5-phophatase; type I inositol-1,4,5-trisphosphate 5-phosphatase
Gene Symbols INPP5A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INPP5A. Recombinant protein control fragment (Product #RP-102466).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3632
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.