Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INPP5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522231
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522231 Supplier Invitrogen™ Supplier No. PA522231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (87%), Rat (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cellular calcium signaling is controlled by the production of inositol phosphates (IPs) by phospholipase C in response to extracellular signals. The IP signaling molecules are inactivated by a family of inositol polyphosphate-5-phosphatases (5-phosphatases). INPP5B encodes the type II 5-phosphatase. The protein is localized to the cytosol and mitochondria, and associates with membranes through an isoprenyl modification near the C-terminus. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP5B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene INPP5B
Gene Accession No. P32019
Gene Alias 5PTase; 75 kDa inositol polyphosphate-5-phosphatase; AW260155; inositol polyphosphate-5-phosphatase B; inositol polyphosphate-5-phosphatase, 75kDa; INPP5B; INPP5P; LOW QUALITY PROTEIN: type II inositol 1,4,5-trisphosphate 5-phosphatase; MGC65156; MGC71303; OCRL2; Phosphoinositide 5-phosphatase; RP11-109P14.7; type II inositol 1,4,5-trisphosphate 5-phosphatase; type II inositol 1,4,5-trisphosphate 5-phosphatase; LOW QUALITY PROTEIN: type II inositol 1,4,5-trisphosphate 5-phosphatase; type II inositol-1
Gene Symbols INPP5B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human INPP5B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3633
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.