Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INPP5E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564248 Supplier Invitrogen™ Supplier No. PA564248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKDTYDSTSK QRTPSYTDRV LYRSRHKGDI CPVSYSSCPG IKTSDHRPVY GLFRVKVRPG RDNIPLAAGK FDRELYLLGI KRRISKEIQR QQALQSQNSS TICSVS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

INPP5E (inositol polyphosphate 5-phosphatase), also known as phosphatidylinositol polyphosphate 5-phosphatase type IV, is a 644 amino acid peripheral membrane protein associated with Golgi stacks. Belonging to the inositol-1, 4, 5-trisphosphate 5-phosphatase type IV family, INPP5E converts phosphatidylinositol-3, 4, 5-triphosphate (PtdIns 3, 4, 5-P3) to PtdIns-P2. While inactive towards water soluble inositol phosphates, the activity of INPP5E is specific for lipid substrates. INPP5E becomes phosphorylated upon DNA damage and is expressed in brain, heart, pancreas, testis and spleen. Defects in INPP5E are the cause of Joubert syndrome type 1 (JBTS1), a disorder presenting with cerebellar ataxia, oculomotor apraxia, hypotonia, neonatal breathing abnormalities and psychomotor delay. In addition, mutations in the INPP5E gene may lead to mental retardation-truncal obesity-retinal dystrophy-micropenis (MORMS), an autosomal recessive disorder characterized by moderate mental retardation, truncal obesity, congenital non-progressive retinal dystrophy, and micropenis in males.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP5E
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Inpp5e
Gene Accession No. Q9NRR6
Gene Alias 1200002L24Rik; 5-phosphatase that induces arborization; 72 kDa inositol polyphosphate 5-phosphatase; 72kDa; CORS1; CPD4; inositol polyphosphate 5-phosphatase; inositol polyphosphate-5-phosphatase E; inositol polyphosphate-5-phosphatase, 72 kDa; INPP5E; JBTS1; mKIAA0123; MORMS; Pharbin; phosphatidylinositol 4,5-bisphosphate 5-phosphatase; Phosphatidylinositol polyphosphate 5-phosphatase type IV; Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase; phosphatidylinositol-4,5-bisphosphate 5-phosphatase; PPI5PIV
Gene Symbols Inpp5e
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INPP5E. Recombinant protein control fragment (Product #RP-103817).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56623
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.