Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INPP5F Monoclonal Antibody (OTI1D4)
GREENER_CHOICE

Catalog No. PIMA527229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527229 Supplier Invitrogen™ Supplier No. MA527229
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

INPP5F Monoclonal Antibody for Western Blot, IHC (P)

SAC2 is an inositol 1,4,5-trisphosphate (InsP3) 5-phosphatase and contains a Sac domain. The activity of this protein is specific for phosphatidylinositol 4,5-bisphosphate and phosphatidylinositol 3,4,5-trisphosphate. Alternatively spliced transcript variants have been observed, but most of them are not thought to be protein-coding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP5F
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene INPP5F
Gene Accession No. Q9Y2H2
Gene Alias 5830435P03Rik; AI115354; AW561896; cI-27; EC 3.1.3.36; hSAC2; Inositol polyphosphate 5-phosphatase F; inositol polyphosphate-5-phosphatase F; INPP5F; Kiaa0966; mKIAA0966; MSTP007; MSTP047; MSTPO47; OCRL 1; phosphatidylinositide phosphatase SAC2; Sac domain-containing inositol phosphatase 2; sac domain-containing phosphoinositide 4-phosphatase 2; sac domain-containing phosphoinositide 5-phosphatase 2; SAC2
Gene Symbols INPP5F
Host Species Mouse
Immunogen Full length human recombinant protein of INPP5F produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22876
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.