Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INPPL1 Rabbit anti-Human, APC, Clone: 1D2, Abnova™

Catalog No. 89335904 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-904 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-904 Supplier Abnova Corporation Supplier No. MAB19070
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

INPPL1 Monoclonal antibody specifically detects INPPL1 in Human samples. It is validated for Flow Cyt.

Specifications

Antigen INPPL1
Applications Flow Cytometry
Classification Monoclonal
Clone 1D2
Conjugate APC
Description inositol polyphosphate phosphatase-like 1
Gene INPPL1
Gene Alias SHIP2
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Y1135 of human INPPL1
Quantity 10 Reactions
Primary or Secondary Primary
Gene ID (Entrez) 3636
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.