Learn More
INPPL1/SHIP2 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 or Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat
Specifications
Specifications
| Antigen | INPPL1/SHIP2 |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | INPPL1 |
| Gene Accession No. | O15357, Q6P549 |
| Gene Alias | 51C protein; INPPL-1; SH2 domain-containing inositol-5′-phosphatase 2; SHIP-2; phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 2; protein 51C |
| Gene Symbols | INPPL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.