Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INSC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558921
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558921 Supplier Invitrogen™ Supplier No. PA558921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CLQVENEHVL KSMKACVSET LSMLGQHFGQ LLELALTREV QALVRKIDAS DNIYTTESTT GNLFSLTQEG APLCRIIAKE GGVVALFKVC RQDSFRCLY Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

In Drosophila, neuroblasts divide asymmetrically into another neuroblast at the apical side and a smaller ganglion mother cell on the basal side. Cell polarization is precisely regulated by 2 apically localized multiprotein signaling complexes that are tethered by Inscuteable, which regulates their apical localization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INSC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Insc
Gene Accession No. Q1MX18
Gene Alias 3830422K02Rik; C730020D23; INSC; inscuteable homolog (Drosophila); inscuteable spindle orientation adaptor protein; minsc; Protein inscuteable homolog; RGD1311379
Gene Symbols Insc
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INSC. Recombinant protein control fragment (Product #RP-97541).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387755
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.