Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insect Cathepsin L Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2259SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2259SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2259SP Supplier R&D Systems Supplier No. MAB2259SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Cathepsin L Monoclonal specifically detects Cathepsin L in Insect samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Cathepsin L
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 193720
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias cathepsin L, cathepsin L1, CATL, CTSL, CTSL1, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP
Gene Symbols CTSL1
Host Species Mouse
Immunogen S. frugiperda ovarian cell line Sf 21-derived Insect Cathepsin L
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1514
Test Specificity Detects pro form of Insect Cathepsin L in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) Cathepsin S is observed and no cross-reactivity with rhCathepsin B, F, K, L2/V, and activated, mature insect Cathepsin L is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Insect
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.