Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insect Cathepsin L Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
$200.50 - $526.00
Specifications
Antigen | Cathepsin L |
---|---|
Clone | 193702 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB22591SP
![]() |
R&D Systems
MAB22591SP |
25 μg |
Each for $200.50
|
|
|||||
MAB22591
![]() |
R&D Systems
MAB22591 |
100 μg |
Each for $526.00
|
|
|||||
Description
Cathepsin L Monoclonal specifically detects Cathepsin L in Insect samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cathepsin L | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Insect | |
cathepsin L, cathepsin L1, CATL, CTSL, CTSL1, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP | |
CTSL1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
193702 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
1514 | |
S. frugiperda ovarian cell line Sf 21-derived Insect Cathepsin L | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects pro and mature forms of Insect Cathepsin L in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Cathepsin L is observed and no cross-reactivity with recombinant mouse Cathepsin L is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title