Learn More
Fisherbrand™ Insert Adapter
Catalog No. 23900181 Shop All Drucker Diagnostics Products
Click to view available options
Capacity:
11 mL
Accessory for use with centrifuges.
  • Refer to centrifuge manual for compatibility confirmation
Specifications
Type Centrifuge Tube Adapter
Tube Type Standard
Material Plastic
Capacity 11 mL
Autoclavable Not Autoclavable
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.