Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter Glass Limited Volume Inserts for 9 mm Autosampler Vials, Untreated
SDP

Catalog No. 15111004 Shop All DWK Life Sciences Products
Click to view available options
No. per Pack:
100
1000

MicroLiter limited volume inserts are made from Type 1 borosilicate glass or polypropylene and meet strict tolerances. Conical inserts, inserts with plastic springs, and flat bottom inserts are available to assist in consistent sampling of low-volume samples.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Borosilicate Glass (Type 1)
No. per Pack 1000
Volume (Metric) Insert 350 μL
Base Style Conical with Pulled Point
Product Line MicroLiter
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.