Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INSIG-1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11055244AF350
Description
INSIG-1 Polyclonal specifically detects INSIG-1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
INSIG-1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405 | |
Rabbit | |
Affinity Purified | |
Cholesterol Metabolism, ER Markers, Lipid and Metabolism, Signal Transduction | |
3638.0 | |
Human, Rat | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
INSIG1 | |
A synthetic peptide representing an internal region of the human INSIG-1 protein (between residues 1-100) according to Swiss-ProtNo. O15503. | |
0.1 mL | |
Primary | |
This is specific for Insig1 but not Insig2. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction