Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INSIG-2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | INSIG-2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
INSIG-2 Polyclonal specifically detects INSIG-2 in Human, Mouse samples. It is validated for Western Blot.Specifications
INSIG-2 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
51141 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
INSIG-2, INSIG2 membrane protein, insulin induced gene 2, insulin induced protein 2, insulin-induced gene 2 protein, MGC26273 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human INSIG-2 (NP_057217.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title