Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INSIG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109862
Description
Immunogen sequence: LWWTFDRSRS GLGLGITIAF LATLITQFLV YNGVYQYTSP DFLYIRSWLP CIFFSGGVTV GNIGRQLAMG VPEKPHSD.
Oxysterols regulate cholesterol homeostasis through liver X receptor (LXR) and sterol regulatory element-binding protein (SREBP) mediated signaling pathway. This gene is an insulin-induced gene. It encodes an endoplasmic reticulum (ER) membrane protein that plays a critical role in regulating cholesterol concentrations in cells. This protein binds to the sterol-sensing domains of SREBP cleavage-activating protein (SCAP) and HMG CoA reductase, and is essential for the sterol-mediated trafficking of the two proteins. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| INSIG1 | |
| Polyclonal | |
| Unconjugated | |
| INSIG1 | |
| 1810013C12Rik; CL6; CL-6; fb55a03; fi35b10; growth response protein (CL-6); immediate-early protein CL-6; INSIG1; INSIG-1; INSIG-1 membrane protein; insulin induced gene 1; insulin induced protein 1; insulin-induced gene 1 protein; Insulin-induced growth response protein CL-6; MGC1405; wu:fb55a03; wu:fi35b10; zgc:55439 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3638, 64194 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 4.56 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O15503, Q08755 | |
| INSIG1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human INSIG1 (NP_005533.2). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction