Learn More
Invitrogen™ INSIG1 Polyclonal Antibody

Description
This antibody does not cross-react with Insig2. Suggested positive control: HepG2 lysate, antigen standard for INSIG1 (transient overexpression lysate).
Specifications
Specifications
| Antigen | INSIG1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.02 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.1% sodium azide |
| Gene | INSIG1 |
| Gene Accession No. | O15503, Q08755 |
| Gene Alias | 1810013C12Rik; CL6; CL-6; fb55a03; fi35b10; growth response protein (CL-6); immediate-early protein CL-6; INSIG1; INSIG-1; INSIG-1 membrane protein; insulin induced gene 1; insulin induced protein 1; insulin-induced gene 1 protein; Insulin-induced growth response protein CL-6; MGC1405; wu:fb55a03; wu:fi35b10; zgc:55439 |
| Gene Symbols | INSIG1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.