Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INSIG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103441
Description
Antibody detects endogenous levels of total INSIG2.
INSIG2 is highly similar to the protein product encoded by gene INSIG1. Both INSIG1 protein and this protein are endoplasmic reticulum proteins that block the processing of sterol regulatory element binding proteins (SREBPs) by binding to SREBP cleavage-activating protein (SCAP), and thus prevent SCAP from escorting SREBPs to the Golgi.The protein encoded by this gene is highly similar to the protein product encoded by gene INSIG1. Both INSIG1 protein and this protein are endoplasmic reticulum proteins that block the processing of sterol regulatory element binding proteins (SREBPs) by binding to SREBP cleavage-activating protein (SCAP), and thus prevent SCAP from escorting SREBPs to the Golgi.
Specifications
INSIG2 | |
Polyclonal | |
Unconjugated | |
INSIG2 | |
2900053I11Rik; C730043J18Rik; Insig2; INSIG-2; INSIG2 membrane protein; INSIG-2 membrane protein; insulin induced factor 2; insulin induced gene 2; insulin induced protein 2; insulin-induced 2 protein; insulin-induced gene 2 protein; insulin-induced membrane protein 2; MGC26273 | |
Rabbit | |
Affinity chromatography | |
RUO | |
288985, 51141, 72999 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q80UA9, Q91WG1, Q9Y5U4 | |
INSIG2 | |
A synthesized peptide derived from human INSIG2(Accession Q9Y5U4), corresponding to amino acid residues M1-V45. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction