Learn More
Invitrogen™ INSIG2 Polyclonal Antibody

Description
Peptide sequence: MAEGETESPG PKKCGPYISS VTSQSVNLMI RGVVLFFIGV FLALVLNLLQ Sequence homology: Cow: 100%; Dog: 92%; Goat: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 92%; Rabbit: 100%; Rat: 92%; Sheep: 100%.
Specifications
Specifications
| Antigen | INSIG2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | INSIG2 |
| Gene Accession No. | Q9Y5U4 |
| Gene Alias | 2900053I11Rik; C730043J18Rik; Insig2; INSIG-2; INSIG2 membrane protein; INSIG-2 membrane protein; insulin induced factor 2; insulin induced gene 2; insulin induced protein 2; insulin-induced 2 protein; insulin-induced gene 2 protein; insulin-induced membrane protein 2; MGC26273 |
| Gene Symbols | INSIG2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.