Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INSIG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109863 Supplier Invitrogen™ Supplier No. PA5109863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QIQRNVTLFP PDVIASIFSS AWWVPPCCGT ASAVIGLLYP CIDRHLGEPH KFKREWSSVM RCVAVFVGIN HASAKVDFDN NIQLSLTLAA LSIGLWWTFD R.

INSIG2 is highly similar to the protein product encoded by gene INSIG1. Both INSIG1 protein and this protein are endoplasmic reticulum proteins that block the processing of sterol regulatory element binding proteins (SREBPs) by binding to SREBP cleavage-activating protein (SCAP), and thus prevent SCAP from escorting SREBPs to the Golgi.The protein encoded by this gene is highly similar to the protein product encoded by gene INSIG1. Both INSIG1 protein and this protein are endoplasmic reticulum proteins that block the processing of sterol regulatory element binding proteins (SREBPs) by binding to SREBP cleavage-activating protein (SCAP), and thus prevent SCAP from escorting SREBPs to the Golgi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INSIG2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene INSIG2
Gene Accession No. Q80UA9, Q91WG1, Q9Y5U4
Gene Alias 2900053I11Rik; C730043J18Rik; Insig2; INSIG-2; INSIG2 membrane protein; INSIG-2 membrane protein; insulin induced factor 2; insulin induced gene 2; insulin induced protein 2; insulin-induced 2 protein; insulin-induced gene 2 protein; insulin-induced membrane protein 2; MGC26273
Gene Symbols INSIG2
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288985, 51141, 72999
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.