Learn More
Thermo Scientific™ InSight Software
Description
Thermo Scientific™ InSight software tools work with Insight motes and sensors to monitor your cold storage equipment’s temperature energy consumption, and other parameters using Wi-Fi or proprietary wireless protocols.
InSight Analytics:
InSight Analytics, powered by KLATU™, is a system designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:
- Equipment critical parameters
- Electrical use and energy consumption
- Asset utilization
- Environmental conditions
InSight Analytics monitors your laboratory equipment 24 hours a day, seven days a week and alerts you when the thresholds are exceeded. The data from InSight Analytics is run through a number of algorithms utilizing the InSight software. With InSight Analytics, you can determine and compare the assets' performance against industry performance standards.
How InSight Analytics works:
- Data is sent wirelessly from the InSight mote via your Wi-Fi network to the InSight web platform fpr storage.
- Once in the Cloud, the data is analyzed by the system, providing real-time comparison of the laboratory equipment's health to industry averages.
- This data analysis enables you to make knowledge-based asset decisions based on key performance indicators such as product service and support, service vs. buy decisions and remote system health monitoring and trouble-shooting for service providers.
Specifications
Specifications
| Type | Insight Software |
| Description | Subscription credits package 250 credits |
| For Use With (Equipment) | All |
Not available outside North America