Learn More
Thermo Scientific™ InSight Software
Catalog No. 11676395
Change view
Click to view available options
Description:
Subscription credits package 100 credits
Subscription credits package 1000 credits
Subscription credits package 20 credits
Subscription credits package 250 credits
Subscription credits package 50 credits
Subscription credits package upgrade 100 credits
Subscription credits package upgrade 1000 credits
Subscription credits package upgrade 50 credits
Subscription credits package upgrade 550 credits
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11-676-395 -
11-676-387 Subscription credits package 20 credits
11-676-388 Subscription credits package 50 credits
11-676-389 Subscription credits package 100 credits
11-676-390 Subscription credits package 250 credits
11-676-391 -
11-676-392 Subscription credits package 1000 credits
11-676-393 Subscription credits package upgrade 50 credits
11-676-394 Subscription credits package upgrade 100 credits
11-676-396 Subscription credits package upgrade 550 credits
11-676-397 Subscription credits package upgrade 1000 credits
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 11-676-395 Supplier Thermo Scientific™ Supplier No. INSSWUPG0250
Add to Cart
Add to Cart
Thermo Scientific™ InSightLink Premisis Manager works with your WLAN and the InSight Cloud.
Thermo Scientific™ InSight software tools work with Insight motes and sensors to monitor your cold storage equipment’s temperature energy consumption, and other parameters using Wi-Fi or proprietary wireless protocols.
InSight Analytics:
InSight Analytics, powered by KLATU™, is a system designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:
  • Equipment critical parameters
  • Electrical use and energy consumption
  • Asset utilization
  • Environmental conditions
InSight Analytics monitors your laboratory equipment 24 hours a day, seven days a week and alerts you when the thresholds are exceeded. The data from InSight Analytics is run through a number of algorithms utilizing the InSight software. With InSight Analytics, you can determine and compare the assets' performance against industry performance standards.
How InSight Analytics works:
  • Data is sent wirelessly from the InSight mote via your Wi-Fi network to the InSight web platform fpr storage.
  • Once in the Cloud, the data is analyzed by the system, providing real-time comparison of the laboratory equipment's health to industry averages.
  • This data analysis enables you to make knowledge-based asset decisions based on key performance indicators such as product service and support, service vs. buy decisions and remote system health monitoring and trouble-shooting for service providers.
TRUSTED_SUSTAINABILITY
Specifications
Type Insight Software
For Use With (Equipment) All
Not available outside North America