Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INSL3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981249
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1249 Supplier Bioss Supplier No. BS11301R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Seems to play a role in testicular function. May be a trophic hormone with a role in testicular descent in fetal life. Is a ligand for LGR8 receptor.

Specifications

Antigen INSL3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene INSL3
Gene Accession No. P51460
Gene Alias INSL 3; INSL3; insulin like 3; Insulin-like 3; insulin-like 3 (Leydig cell); Insulin-like 3 A chain; Insulin-like 3 B chain; Ley IL; leydig insulin -like hormone; Leydig insulin-like peptide; Ley-I-L; prepro-INSL3; Relaxin like factor; Relaxin-like factor; relaxin-like factor b; RLF; RLNL
Gene Symbols INSL3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human INSL3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3640
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.