Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INSL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106660 Supplier Invitrogen™ Supplier No. PA5106660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total INSL4.

Insulin is a secreted peptide hormone that elicits metabolic effects such as increases in glucose uptake and glycogen synthesis leading to a decrease in blood glucose concentration. Insulin is first formed as a precursor molecule, preproinsulin, which is later cleaved to proinsulin and finally to the mature insulin hormone. Insulin-like peptides (INSL proteins), also designated Relaxinlike factors, are members of the insulin family, which regulate cell growth, metabolism and tissue-specific functions. INSL1-7 are mostly secreted proteins that are expressed mainly in testis, placenta, uterus or prenatal tissues. INSL4 is a 139 amino acid secreted protein expressed in the placenta, uterus and in fetal perichondrium. It may play an important role in the regulation of bone formation and in trophoblast development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INSL4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene INSL4
Gene Accession No. Q14641
Gene Alias early placenta insulin-like peptide; early placenta insulin-like peptide (EPIL); Early placenta insulin-like peptide A chain; Early placenta insulin-like peptide B chain; EPIL; INSL4; insulin like 4; insulin-like 4 (placenta); insulin-like peptide 4; Placentin
Gene Symbols INSL4
Host Species Rabbit
Immunogen A synthesized peptide derived from human INSL4(Accession Q14641), corresponding to amino acid residues K48-S98.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3641
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.