Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INSL4 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304443100UL
Description
INSL4 Polyclonal specifically detects INSL4 in Rat samples. It is validated for Western Blot.Specifications
INSL4 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
3641 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
early placenta insulin-like peptide, EPILearly placenta insulin-like peptide (EPIL), insulin-like 4 (placenta), Insulin-like peptide 4, Placentin | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-139 of human INSL4 (NP_002186.1). | |
100 μL | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction