Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INSL5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543766
Description
Peptide sequence: RTVIYICASS RWRRHQEGIP QAQQAETGNS FQLPHKREFS EENPAQNLPK Sequence homology: Human: 100%; Rabbit: 79%.
The protein encoded by this gene contains a classical signature of the insulin superfamily and is highly similar to relaxin 3 (RLN3/INSL7).
Specifications
INSL5 | |
Polyclonal | |
Unconjugated | |
INSL5 | |
Insl5; insulin like 5; insulin-like 5; insulin-like peptide 5; Insulin-like peptide INSL5; Insulin-like peptide INSL5 A chain; Insulin-like peptide INSL5 B chain; prepro-INSL5; PRO182; Relaxin/insulin-like factor 2; relaxin/insulin-like protein; Rif; Rif2; UNQ156; UNQ156/PRO182; Zins3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10022 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y5Q6 | |
INSL5 | |
Synthetic peptide directed towards the middle region of human INSL5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction