Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INSL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556342
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556342 Supplier Invitrogen™ Supplier No. PA556342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEYIRTVIYI CASSRWRRHL EGIPQAQQAE TGNSFQLPHK REFSEENPAQ NLPKVDASGE DRLWGGQMPT EELWKSKKHS VMSR Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 29%.

The protein encoded by this gene contains a classical signature of the insulin superfamily and is highly similar to relaxin 3 (RLN3/INSL7).
TRUSTED_SUSTAINABILITY

Specifications

Antigen INSL5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene INSL5
Gene Accession No. Q9Y5Q6
Gene Alias Insl5; insulin like 5; insulin-like 5; insulin-like peptide 5; Insulin-like peptide INSL5; Insulin-like peptide INSL5 A chain; Insulin-like peptide INSL5 B chain; prepro-INSL5; PRO182; Relaxin/insulin-like factor 2; relaxin/insulin-like protein; Rif; Rif2; UNQ156; UNQ156/PRO182; Zins3
Gene Symbols INSL5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INSL5. Recombinant protein control fragment (Product #RP-94456).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10022
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.