Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INSL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106458
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106458 Supplier Invitrogen™ Supplier No. PA5106458
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total INSL6.

The protein encoded by this gene contains a classical signature of the insulin superfamily and is significantly similar to relaxin and relaxin-like factor. This gene is preferentially expressed in testis. Its expression in testis is restricted to interstitial cells surrounding seminiferous tubules, which suggests a role in sperm development and fertilization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INSL6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene INSL6
Gene Accession No. Q9QY05, Q9Y581
Gene Alias INSL6; insulin like 6; insulin-like 6; insulin-like peptide 5; insulin-like peptide 6; Insulin-like peptide INSL6; Insulin-like peptide INSL6 A chain; Insulin-like peptide INSL6 B chain; Relaxin/insulin-like factor 1; RIF1
Gene Symbols INSL6
Host Species Rabbit
Immunogen A synthesized peptide derived from human INSL6(Accession Q9Y581), corresponding to amino acid residues N95-A145.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11172, 27356
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.