Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INSM2 Mouse anti-Human, Clone: INSM2/4291, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
| Antigen | INSM2 |
|---|---|
| Clone | INSM2/4291 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
INSM2 Monoclonal specifically detects INSM2 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| INSM2 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| insulinoma-associated 2, Nbla106, zinc finger protein IA-6 | |
| Recombinant fragment of human INSM2 protein (around aa 432-540) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| INSM2/4291 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 84684 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title