Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INSR alpha Chimeric Recombinant Rabbit Monoclonal Antibody (83-14)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547791
Description
Specificity: Recognises the extracellular domain of the Insulin Receptor alpha. Amino acids 469-592 (exon 7 and 8) of Insulin Receptor alpha.
Biological actions of insulin and IGF-1 are mediated by their respective cell surface receptor tyrosine kinases that regulate multiple signaling pathways through activation of a series of phosphorylation cascades. The insulin receptor. Insulin/IGF-1 binding to the extracellular domain leads to autophosphorylation of downstream target proteins. These two receptors differ in sequence in regions that confer specificity for the designated ligand as well as in certain intracellular signaling domains, resulting in significant differences in the functional consequences of activation of each receptor. Defects in IR are the cause of various insulin resistance syndromes and IGF-1R defects may cause some forms of growth retardation. Both these signaling cascades are also important for the development of cancer.
Specifications
| INSR alpha Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P06213 | |
| INSR | |
| This antibody was raised by immunizing mice with IM-9 lymphocytes followed by purified insulin receptor. | |
| 200 μg | |
| Primary | |
| Human, Bovine, Sheep, Pig | |
| Antibody | |
| IgG κ |
| ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 83-14 | |
| Unconjugated | |
| INSR | |
| 4932439J01Rik; CD220; CD220 beta; D630014A15Rik; DAF-2; HHF5; INSR; insulin receptor; insulin receptor (INSR); insulin receptor precursor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR alpha; IR beta; ir-A; IR-B | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3643, 396755, 408017, 443431 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction