Learn More
Promega InsR Kinase Enzyme System, 1mg
Catalog No. PRV3902 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3902 1 mg
1 options
Catalog No. PRV3902 Supplier Promega Supplier No. V3902
Add to Cart
Add to Cart
Request a Quote
Recombinant human InsR (amino acids 1011-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. InsR is the insulin receptor tyrosine kinase that is involved in insulin signaling.
  • Easily Screen and Profile InsR Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C