Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INSRR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5120068 Supplier Invitrogen™ Supplier No. PA5120068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive test controls include: Mouse brain, Mouse liver, Mouse kidney, Rat kidney. The target is usually found in the following locations: Membrane, Single-pass type I membrane protein. Immunogen sequence: AAGPLRLGGN SSDFEIQEDK VPRERAVLSG LRHFTEYRID IHACNHAAHT VGCSAATFVF ARTMPHREAD GIPGKVAWEA SSKNSVLLRW LEPPDPNGLI LKYEIKYRRL GEEATVLCVS RLRYAKFGGV HLALLPPGNY SARVRATSLA GNGSWTDSVA FYILGPEEED AGGLH.

INSRR, insulin receptor-related receptor, is a member of the insulin receptor (IR) family. INSRR does not bind insulin-like molecules, but is similar to INSR in tyrosine kinase activity. Receptor with tyrosine-protein kinase activity. Functions as a pH sensing receptor which is activated by increased extracellular pH. Activates an intracellular signaling pathway that involves IRS1 and AKT1/PKB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INSRR
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Insrr
Gene Accession No. Q64716, Q9WTL4
Gene Alias Insrr; insulin receptor related receptor; insulin receptor-related protein; Insulin receptor-related protein alpha chain; Insulin receptor-related protein beta chain; insulin receptor-related receptor; IRR; IR-related receptor; Sirr
Gene Symbols Insrr
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 747-921 of human INSRR (NP_055030.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23920, 60663
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.