Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INSRR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110908
Description
Immunogen sequence: LCLEHIYRLE EVTGTRGRQN KAEINPRTNG DRAACQTRTL RFVSNVTEAD RILLRWERYE PLEARDLLSL IVYYKESPFQ NATEHVGPDA CGTQSWNLLD VELPLSRTQE PGVTLASLKP WTQYAVFVRA ITLTTEEDSP HQGAQ.
INSRR, insulin receptor-related receptor, is a member of the insulin receptor (IR) family. INSRR does not bind insulin-like molecules, but is similar to INSR in tyrosine kinase activity. Receptor with tyrosine-protein kinase activity. Functions as a pH sensing receptor which is activated by increased extracellular pH. Activates an intracellular signaling pathway that involves IRS1 and AKT1/PKB.
Specifications
| INSRR | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P14616 | |
| Insrr | |
| Recombinant Protein corresonding to Human INSRR. Recombinant protein control fragment (Product #RP-89147). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Insrr | |
| Insrr; insulin receptor related receptor; insulin receptor-related protein; Insulin receptor-related protein alpha chain; Insulin receptor-related protein beta chain; insulin receptor-related receptor; IRR; IR-related receptor; Sirr | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3645 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction