Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ INSTA-Blot Human Tissues
SDP

Catalog No. NB397866 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Membrane
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397866 1 Membrane
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397866 Supplier R&D Systems™ Supplier No. NBP311812

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

INSTA-Blot™ is a ready-to-use PVDF membrane containing denatured protein from human tissue lysates, designed to streamline the western blotting procedure by eliminating sample acquisition, preparation, SDS-PAGE, and electro-blotting steps.

  • Applications: Western Blot
  • Species Reactivity: Human
  • Storage: Store at room temperature
  • Guarantee: 1 Year from date of receipt
  • Buffer Composition: Tissues are lysed in a buffer containing 10 mM Tris (pH 8.0), 130 mM NaCl, 1% Triton X-100, 10 mM NaF, 10 mM NaPi, 10 mM NaPPi, Protease Inhibitor Cocktail (PIC), and PMSF. Lysates are resuspended in 2X SDS sample buffer containing 120 mM Tris-HCl (pH 6.8), 20 mM EDTA, 4% SDS, 0.06% Bromophenol Blue, 20% glycerol, and 0.4% β-mercaptoethanol.

Specifications

For Use With (Application) Western Blot
Quantity 1 Membrane
Content And Storage Store at room temperature.
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.