Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips



Click to view available options
Parameter Specifications:
Chlorine - free, high range
Chlorine - free, low range
Chlorine - total, low range
Chlorine dioxide
Copper
Peracetic Acid
Sodium Chloride
Quantity:
10 Papers/Blister Card, 10 Cards/Pk.
25 Strips/Vial
50 Strips/Vial
Range:
0 to 10 ppm
0 to 10 ppm (0, 0.25, 0.5, 1, 3, 10)
0 to 50 ppm
0 to 500 ppm
0-0.3-0.6-1-3 ppm
0-0.5-1-3-5-10 ppm
0-50-100-250-500-800 ppm
1500 to 5000 ppm
Description
- No reagents or field kits required for most strips
- Chemically treated pads on plastic strips produce color change
- Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
- As little as 10-second development time
- Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †
Specifications
Specifications
For Use With (Application) | Water quality testing |
Parameter Specifications | Sodium Chloride |
Quantity | 10 Papers/Blister Card, 10 Cards/Pk. |
Range | 1500 to 5000 ppm |
Storage Requirements | Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight |
Type | Single Factor Test Strip |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction