Learn More
Description
- Monitor water without having to use reagents or field kits.
Order Info
Each vial of test strips contains 50 tests per factor, except S66233, which contains 25 tests per factor.
Specifications
Specifications
| Analytes | Hardness |
| Format | Strip |
| For Use With (Application) | Drinking water, food/beverage testing |
| Quantity | 50 Pk. |
| Range | 0 to 180 ppm (0, 30, 60, 120, 180 ppm) |
| Type | Water Test Strip |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

