Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Instant Light Powder Chemiluminescence Demo Kit
SCI_ED

Catalog No. s06632
Click to view available options
Class Size:
5 Demonstrations

Includes

One bottle instant-light powder

Designed to help students understand the topics of chemiluminescence and kinetics.

  • Instant-light powder and water produces blue light when placed in a beaker
  • For five demonstrations

Specifications

Product Type Chemiluminescence Demonstration Kit
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Designed to help students understand the topics of chemiluminescence and kinetics
Includes One bottle instant-light powder
Quantity 1
Dimensions (L x W x H) 4.25 x 4.625 x 5.5 in.
Class Size 5 Demonstrations
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.