Learn More
Description
- Help protect ion selective electrodes (ISE) and eliminate additional caps, tubes and specimen labeling
- Use with glass or plastic collection tubes
- Technique reduces second centrifugation, tube labeling and sample preparation time
- Allow direct sampling into automatic laboratory equipment
- Eliminate fibrin, particulate matter and equipment downtime
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Diameter (Metric) Outside | 16mm |
| Diameter (English) Outside | 0.6 in. |
| Length (English) | 1.87 in. |
| For Use With (Application) | Remove fibrin and gel separation particulates and permit direct sampling from primary collection tubes |
| Quantity | 100 Pack |
| Product Type | Serum Filter |
| For Use With (Equipment) | Collection tube 16 x 100mm |
| Length (Metric) | 47mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
