Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin Antibody (01), Novus Biologicals™

Mouse Monoclonal Antibody
$322.50
Specifications
Antigen | Insulin |
---|---|
Clone | 1 |
Dilution | ELISA 1:1000-1:2000, Sandwich ELISA Capture 1:250-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Insulin Monoclonal antibody specifically detects Insulin in Human samples. It is validated for ELISASpecifications
Insulin | |
ELISA 1:1000-1:2000, Sandwich ELISA Capture 1:250-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Insulin (Uniprot#: P01308; Phe25-Lys53 & Gly90-Asn110). | |
Protein A purified |
1 | |
ELISA | |
Unconjugated | |
Mouse | |
Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers | |
0.2 um filtered solution in PBS | |
3630 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title