Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Antibody (D6C4cc), Novus Biologicals™
SDP

Catalog No. NB1208304 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1208304 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1208304 Supplier Novus Biologicals Supplier No. NB1208304
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Insulin Monoclonal specifically detects Insulin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen, Sandwich ELISA.

Specifications

Antigen Insulin
Applications Immunohistochemistry, Immunohistochemistry (Frozen), ELISA
Classification Monoclonal
Clone D6C4cc
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Sandwich ELISA 1:100-1:2000
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Gene Symbols INS
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with native human insulin.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3630
Test Specificity This is specific for insulin and proinsulin.
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.