Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin Antibody (IRDN/1980R), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308836AF350
Description
Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse, Porcine, Bovine, Rabbit samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Insulin | |
Monoclonal | |
Alexa Fluor 350 | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
Recombinant human full-length Insulin protein (IRDN) (Uniprot: P01308) | |
100 μg | |
Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers | |
3630 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
IRDN/1980R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Pig, Bovine, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction