Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Insulin Monoclonal Antibody (ICBTACLS), eBioscience™

Catalog No. 14976937
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-976-937 2 mg
50-112-2850 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-976-937 Supplier Invitrogen™ Supplier No. 14976937
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This ICBTACLS monoclonal antibody reacts with the hormone insulin, responsible for regulation of carbohydrate and fat metabolism. Blood glucose levels are maintained by insulin. Insulin secretion from pancreatic beta-cells within the islets of Langerhans results in uptake of excess glucose from the blood into organs and its storage in the form of glycogen. Patients with type 1 diabetes have an autoimmune destruction of insulin-producing beta-cells in the pancreas, resulting in insulin deficiency and elevated levels of blood glucose. Patients with type 2 diabetes have insulin resistance where insulin becomes less effective at reducing the amount of blood glucose. Insulin is also important for proper endothelial cell function in with respect to vascular elasticity in response to changing blood pressure and plays a role in cognition, learning, and memory. The ICBTACLS antibody recognizes human, mouse, and rat insulin. Applications Reported: This ICBTACLS antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and microscopy. Applications Tested: This ICBTACLS antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using low or high pH antigen retrieval and can be used at less than or equal to 0.25 μg/mL. This ICBTACLS antibody has been tested by western blot of non-reduced pancreatic tissue lysate and can be used at less than or equal to 5 ...

Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Insulin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone ICBTACLS
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene INS
Gene Accession No. P01308, P01322, P01323, P01325, P01326
Gene Alias AA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2; Ins-2; Ins2-rs1; InsII; Insulin; insulin 1; insulin 2; Insulin A chain; Insulin B chain; insulin I; insulin II; insulin-1; Insulin-1 A chain; Insulin-1 B chain; Insulin-2; Insulin-2 A chain; Insulin-2 B chain; IRDN; Mody; MODY10; Mody4; preproinsulin; proinsulin
Gene Symbols INS, Ins1, Ins2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16333, 16334, 24505, 24506, 3630
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.