Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin/Proinsulin Antibody (D3E7cc), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB200435UV
Description
Insulin/Proinsulin Monoclonal antibody specifically detects Insulin/Proinsulin in Human, Rat, Porcine, Bovine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), ELISASpecifications
Insulin/Proinsulin | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Rat, Pig, Bovine | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), ELISA | |
D3E7cc | |
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Sandwich ELISA | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with native human insulin. | |
0.2 mL | |
Diabetes Research | |
3630 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction