Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin R/CD220 Antibody (4B2), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326192100UL
Description
Insulin R/CD220 Monoclonal antibody specifically detects Insulin R/CD220 in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
Insulin R/CD220 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunocytochemistry | |
4B2 | |
ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
A synthesized peptide derived from Human Insulin R/CD220 [UniProt P06213] | |
100 μL | |
Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
3643 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction