Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin R/CD220 Antibody (INSR/2277R), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308838FR
Description
Insulin R/CD220 Monoclonal antibody specifically detects Insulin R/CD220 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Insulin R/CD220 | |
Monoclonal | |
DyLight 680 | |
CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
Recombinant fragment of extracellular domain of human Insulin Receptor alpha (exact sequence is proprietary) (Uniprot: P06213) | |
100 μg | |
Diabetes Research, Lipid and Metabolism, Neuroscience, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
3643 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
INSR/2277R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction