Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin Receptor beta, p Thr1160 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260777
Description
Insulin Receptor beta Polyclonal specifically detects Insulin Receptor beta in Human, Mouse, Rat, Porcine, Bovine, Canine, Primate, Xenopus samples. It is validated for Western Blot.Specifications
Insulin Receptor beta (p Thr1160) | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
Rabbit | |
95 kDa | |
100 μL | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Canine, Primate, Xenopus species | |
IgG |
Western Blot | |
Unconjugated | |
10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. with No Preservative | |
INSR | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr1160 conjugated to KLH. Accession # P06213 | |
Antigen Affinity-purified | |
Lipid and Metabolism, Neuroscience, Nutrient Sensing in the Brain, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
3643 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction