Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulysin/IDE Antibody (334501), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2496MFV450
Description
Insulysin/IDE Monoclonal antibody specifically detects Insulysin/IDE in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| Insulysin/IDE | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 3416 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, CyTOF | |
| 334501 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Abeta-degrading protease, EC 3.4.24, EC 3.4.24.56, FLJ35968, Insulin protease, insulinase, insulin-degrading enzyme, Insulysin | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Insulysin/IDE, Met42-Leu1019, Accession # P14735 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction