Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulysin/IDE Rabbit anti-Human, Mouse, Rat, Zebrafish, Clone: JJ0949, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP266907
Description
Insulysin/IDE Monoclonal specifically detects Insulysin/IDE in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Insulysin/IDE | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
Abeta-degrading protease, EC 3.4.24, EC 3.4.24.56, FLJ35968, Insulin protease, insulinase, insulin-degrading enzyme, Insulysin | |
Rabbit | |
Protein A purified | |
Lipid and Metabolism | |
3416 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
JJ0949 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
IDE | |
Recombinant protein within Human Insulysin/IDE aa 1-106/ 1019. (SwissProt: P14735 Human; SwissProt: Q9JHR7 Mouse; SwissProt: P35559 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction