Learn More
INT4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00311

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Chicken, Turkey, Rabbit, Guinea pig_10141, Pig, Orangutan, Monkey, Gorilla, Ch
INTS4 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1 (RNU1; MIM 180680) and U2 (RNU2; MIM 180690) (Baillat et al., 2005 [PubMed 16239144]).Specifications
INT4 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q96HW7 | |
INTS4 | |
Between 913 and 963 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
INTS4 | |
INT4, MST093, MSTP093 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92105 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.