Learn More
INT7 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00413T

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Chicken, Turkey, Bovine, Dog, Horse, Pig, Panda, Orangutan, Monkey, Gorilla, Chimpanzee and North
Integrator complex subunit 7 (INT7) is a component of the Integrator complex. The Integrator complex is a complex associates with the C-terminal domain of RNA polymerase II and mediates 3′-end processing of the small nuclear RNAs U1 and U2. It plays a role in DNA damage response signaling during the S phase.Specifications
INT7 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9NVH2 | |
INTS7 | |
Between 850 and 900 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
INTS7 | |
C1orf73, chromosome 1 open reading frame 73, DKFZP434B168, INT7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25896 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.