Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INT7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584902
Description
Immunogen sequence: GDGMLGDLME LYKVIGRSAT DKQQELLVSL ATVIFVASQK ALSVESKAVI KQQLESVSNG WTVYRIARQA SRMGNHDMAK ELYQSLLTQV A.
This gene encodes a subunit of the integrator complex. The integrator complex associates with the C-terminal domain of RNA polymerase II and mediates 3'-end processing of the small nuclear RNAs U1 and U2. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
INT7 | |
Polyclonal | |
Unconjugated | |
INTS7 | |
5930412E23Rik; C1orf73; DKFZP434B168; Int7; Integrator complex subunit 7; Ints7; RGD1308014 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25896 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NVH2 | |
INTS7 | |
Recombinant protein corresponding to Human INTS7. Recombinant protein control fragment (Product #RP-108599). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction