Learn More
Integra™ Miltex™ Jacobson Microvascular Needle Holders
Catalog No. p-4185170
Change view
Click to view available options
Length (Metric):
17.8 cm
21.6 cm
Style:
Serrated, curved tip
Serrated, straight tip
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Style
12-460-223 17.8 cm Serrated, straight tip
12-460-224 17.8 cm Serrated, curved tip
12-460-225 21.6 cm Serrated, straight tip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-460-223 Supplier Integra™ Miltex™ Supplier No. 171090TC
Only null left
Add to Cart
Add to Cart
Locking
  • Serrated jaws
  • 16,000 teeth/sq. in.
Specifications
Product Type Microvascular Needle Holder
Material Tungsten Carbide
Includes Lock
Style Serrated, straight tip
Teeth Count 16000 Teeth/sq. in.
Length (English) 7 in.
Length (Metric) 17.8 cm